MES lysis buffer with Triton® X-100, Thermo Scientific

Supplier: Thermo Scientific Chemicals

J62693-AK J62693-AP
CAAAJ62693-AKEA 106.79 CAD
CAAAJ62693-AK CAAAJ62693-AP
MES lysis buffer with Triton® X-100, Thermo Scientific
Buffers Extraction Buffers

Liquid


Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Appearance: Colorless

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR