Phosphate-Buffered Saline (PBS, 10X), pH 7.6, Thermo Scientific

Supplier: Thermo Scientific Chemicals

J62692-K2 J62692-K3 J62692-K7
CAAAJ62692-K2EA 121.41 CAD
CAAAJ62692-K2 CAAAJ62692-K3 CAAAJ62692-K7
Phosphate-Buffered Saline (PBS, 10X), pH 7.6, Thermo Scientific
Buffers Cell Culture Buffers

Liquid


Physical Form: Liquid
MDL No.: MFCD00131855

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR