Cefotaxime sodium salt
Supplier: Thermo Scientific Chemicals
|
Danger
|
J62690-03
J62690-06
J62690-14
CAAAJ62690-03EA
237.93
CAD
CAAAJ62690-03
CAAAJ62690-06
CAAAJ62690-14
Cefotaxime sodium salt
Cefotaxime sodium salt
Powder
CAS No.: 64485-93-4
Molecular Formula: C16H16N5NaO7S2
Formula Weight: 477.40
Physical Form: Powder
Appearance: White to pale yellow
Solubility: Freely soluble in water
Beilstein Reference: 5711411
MDL No.: MFCD00079073
Formula:
C₁₆H₁₇N₅O₇S₂ Storage Temperature: Ambient |
MDL Number:
MFCD00079073 CAS Number: 64485-93-4 EINECS: 264-915-9 Merck Index: 14,1933 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...