Pyridoxamine dihydrochloride cell culture reagent

Supplier: Thermo Scientific Chemicals
Warning

J62679-06 J62679-09 J62679-27
CAAAJ62679-06EA 512.88 CAD
CAAAJ62679-06 CAAAJ62679-09 CAAAJ62679-27
Pyridoxamine dihydrochloride cell culture reagent
Pyridoxamine dihydrochloride

A vitamin in the Vitamin B-6 family


Solid


Applications: A vitamin in the Vitamin B-6 family

Formula: C₈H₁₂N₂O₂·2HCl
Melting Pt: 220…224 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00012808
CAS Number: 524-36-7
EINECS: 208-357-6
Merck Index: 14,7980

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR