G418 disulphate cell culture reagent

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: G418 disulfate, G-418 sulfate, G-418 sulphate, Geneticin®, G-418 disulphate, G418 sulphate, G-418 disulfate

J62671-03 J62671-06
CAAAJ62671-03EA 275.28 CAD
CAAAJ62671-03 CAAAJ62671-06
G418 disulphate cell culture reagent
G418 disulphate

Aminoglycoside that is toxic to bacteria, yeast, protozoans and helminths


Powder


Aminoglycoside that is toxic to bacteria, yeast, protozoans and helminths

Formula: C₂₀H₄₄N₄O₁₈S₂
Storage Temperature: Ambient
MDL Number: MFCD00058314
CAS Number: 108321-42-2

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR