Tolmetin sodium salt dihydrate ≥98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Methyl-5-[p-Toluoyl]Pyrrole-2 Acetic Acid sodium salt dihydrate

J62663-03 J62663-06 J62663-14
CAAAJ62663-03EA 111.82 CAD
CAAAJ62663-03 CAAAJ62663-06 CAAAJ62663-14
Tolmetin sodium salt dihydrate ≥98%
Tolmetin sodium salt dihydrate

An NSAID compound


Powder


Applications: An NSAID compound

Formula: C₁₅H₁₈NNaO₅
MW: 315.301 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00150761
CAS Number: 64490-92-2
EINECS: 252-687-3
Merck Index: 14,9518

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR