TRIS-buffered saline (TBS, 10X) pH 7.6, Thermo Scientific

Supplier: Thermo Scientific Chemicals

J62662-K3 J62662-K7
CAAAJ62662-K3EA 225.1 CAD
CAAAJ62662-K3 CAAAJ62662-K7
TRIS-buffered saline (TBS, 10X) pH 7.6, Thermo Scientific
Buffers Electrophoresis Buffers Washing Buffers

Liquid


CAS No.: 77-86-1
Physical Form: Liquid
MDL No.: MFCD00004679

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR