Norfloxacin

Supplier: Thermo Scientific Chemicals

Synonyms: 1-Ethyl-6-fluoro-1,4-dihydro-4-oxo-7-(1-piperazinyl)-3-quinolinecarboxylic Acid

J62652-03 J62652-06 J62652-14
CAAAJ62652-03EA 51.32 CAD
CAAAJ62652-03 CAAAJ62652-06 CAAAJ62652-14
Norfloxacin
Norfloxacin

A synthetic fluoroquinolone antibiotic


Powder


Applications: A synthetic fluoroquinolone antibiotic

Formula: C₁₆H₁₈FN₃O₃
MW: 319.34 g/mol
Melting Pt: 220…221 °C
Storage Temperature: Ambient
MDL Number: MFCD00079532
CAS Number: 70458-96-7
EINECS: 274-614-4
Merck Index: 14,6700

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR