Neutral red ACS

Supplier: Thermo Scientific Chemicals

Synonyms: N2,N2,7-Trimethylphenazine-2,8-diamine hydrochloride, C.I. 50040, 3-Amino-7-dimethyl amino-2-methylphenazine hydrochloride, N8,N8,3-Trimethyl-2,8-phenazinediamine monohydrochloride, Toluylene red, Basic Red 5

J62643-18 J62643-30
CAAAJ62643-18EA 85.29 CAD
CAAAJ62643-18 CAAAJ62643-30
Neutral red ACS
Neutral red

Nuclear counterstain useful in pH range 6.8 - 8.0.


Powder


Nuclear counterstain useful in pH range 6.8 - 8.0

Formula: C₁₅H₁₇ClN₄
MW: 288.78 g/mol
Melting Pt: 290 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012651
CAS Number: 553-24-2
EINECS: 209-035-8
Merck Index: 14,6488

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR