Stains-all 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-Ethyl-2-[3-(1-ethylnaphtho[1,2-d]thiazolin-2-ylidene)-2-methylpropenyl]naphtho[1,2-d]thiazolium bromide, 3,3′-Diethyl-9-methyl-4,5,4′,5′-dibenzothiacarbocyanine
J62630-03
J62630-06
CAAAJ62630-03EA
143.34
CAD
CAAAJ62630-03
CAAAJ62630-06
Stains-all 95%
Stains-all
Cationic carbocyanine dye used in electrophoresis to stain proteins and nucleic acids
Powder
Applications: Cationic carbocyanine dye used in electrophoresis to stain proteins and nucleic acids
Formula:
C₃₀H₂₇BrN₂S₂ MW: 559.584 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00078788 CAS Number: 7423-31-6 EINECS: 231-047-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...