HEPES-Buffered Saline, pH 7.0 (2X for transfection), Thermo Scientific
Supplier: Thermo Scientific Chemicals
J62623-AK
J62623-AP
CAAAJ62623-AKEA
175.14
CAD
CAAAJ62623-AK
CAAAJ62623-AP
HEPES-Buffered Saline, pH 7.0 (2X for transfection), Thermo Scientific
Buffers
Cell Culture Buffers
Liquid
CAS No.: 7365-45-9
Physical Form: Liquid
MDL No.: MFCD00006158
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...