Valethamate bromide 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
J62622-06
J62622-14
CAAAJ62622-06EA
159.98
CAD
CAAAJ62622-06
CAAAJ62622-14
Valethamate bromide 99%
Valethamate bromide
Powder
Purity: 99%
CAS No.: 90-22-2
Molecular Formula: C19H32BrNO2
Formula Weight: 386.37
Physical Form: Powder
Appearance: White to off-white
Melting Point: 120-127°
MDL No.: MFCD00031617
Formula:
C₁₉H₃₂BrNO₂ Melting Pt: 120…127 °C Storage Temperature: Ambient |
MDL Number:
MFCD00031617 CAS Number: 90-22-2 EINECS: 201-977-8 Merck Index: 14,9906 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...