L-thyroxine 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
(S)-2-Amino-3-(4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl)propanoic acid
J62606-03
J62606-06
CAAAJ62606-03EA
200.98
CAD
CAAAJ62606-03
CAAAJ62606-06
L-thyroxine 98%
L-thyroxine
Crystalline powder
Purity: 98%
CAS No.: 51-48-9
Molecular Formula: C15H11I4NO4
Formula Weight: 776.87
Physical Form: Crystalline powder
Sensitivity: Light Sensitive
Appearance: White to pale brown
Odor: Odorless
Melting Point: 235-236°
Solubility: Dissolves in 4M ammonium hydroxide in Methanol at 50mg/ml
Beilstein Reference: 2228515
MDL No.: MFCD00002595
Formula:
C₁₅H₁₁I₄NO₄ MW: 776.874 g/mol Melting Pt: 235…236 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002595 CAS Number: 51-48-9 EINECS: 200-101-1 Merck Index: 14,9415 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...