Imidazole, 0.5 M Buffer Solution, pH 7.0, Thermo Scientific

Supplier: Thermo Scientific Chemicals

J62593-AE J62593-AK
CAAAJ62593-AEEA 170.01 CAD
CAAAJ62593-AE CAAAJ62593-AK
Imidazole, 0.5 M Buffer Solution, pH 7.0, Thermo Scientific
Buffers Biological Buffers

Liquid


CAS No.: 288-32-4
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
MDL No.: MFCD00005183

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR