Magnesium chloride hexahydrate Cell Culture

Supplier: Thermo Scientific Chemicals

Synonyms: Magnesium dichloride

J62575-30 J62575-36 J62575-A1
CAAAJ62575-30EA 145.28 CAD
CAAAJ62575-30 CAAAJ62575-36 CAAAJ62575-A1
Magnesium chloride hexahydrate Cell Culture
Magnesium chloride hexahydrate

Powder


CAS No.: 7791-18-6 Molecular Formula: MgCl2.6H2O Formula Weight: 203.30 (95.22anhy) Physical Form: Powder Appearance: White Odor: Odorless Merck Reference: 14,5662 MDL No.: MFCD00149781

Formula: MgCl₂·6H₂O
MW: 203.3 g/mol
Boiling Pt: 1412
Melting Pt: 117
Storage Temperature: Ambient
MDL Number: MFCD00149781
CAS Number: 7791-18-6
EINECS: 232-094-6
Merck Index: 14,5662

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR