ß-Nicotinamide adenine dinucleotide phosphate (NADP-Na2, oxidized form) 97%

Supplier: Thermo Scientific Chemicals

Synonyms: Nadide phosphate disodium salt, Triphosphopyridine nucleotide, disodium salt, NADP-Na2, β-NADP-Na2, NADP disodium salt, Nicotinamide-adenine dinucleotide phosphate disodium salt, 1-((2R,3R,4S,5R)-5-((((((((2R,3R,4R,5R)-5-(6-Amino-9H-purin-9-yl)-3-hydroxy-4-(phosphonooxy)tetrahydrofuran-2-yl)methoxy)(hydroxy)phosphoryl)oxy)oxidophosphoryl)oxy)methyl)-3,4-dihydroxytetrahydrofuran-2-yl)-3-carbamoylpyridin-1-ium, disodium salt, β-NADH phosphate disodium salt

J62556-03 J62556-MD
CAAAJ62556-03EA 694.81 CAD
CAAAJ62556-03 CAAAJ62556-MD
ß-Nicotinamide adenine dinucleotide phosphate (NADP-Na2, oxidized form) 97%
ß-Nicotinamide adenine dinucleotide phosphate (NADP-Na2, oxidized form)

Powder


Purity: 97% CAS No.: 24292-60-2 Molecular Formula: C21H26N7Na2O17P3 Formula Weight: 787.37 Storage Temperature: -30°C to -10°C Physical Form: Powder MDL No.: MFCD00065390

Formula: C₂₁H₂₆N₇Na₂O₁₇P₃
MW: 787.37
Storage Temperature: Refrigerator
MDL Number: MFCD00065390
CAS Number: 24292-60-2
EINECS: 246-129-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR