Flumazenil 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Ethyl 8-fluoro-5-methyl-6-oxo-5,6-dihydro-4H-benzo[f]imidazo[1,5-a][1,4]diazepine-3-carboxylate

J62537-MC J62537-ME
CAAAJ62537-MCEA 620.32 CAD
CAAAJ62537-MC CAAAJ62537-ME
Flumazenil 98%
Flumazenil

Benzodiazepine antagonist


Powder


Applications: Benzodiazepine antagonist

Formula: C₁₅H₁₄FN₃O₃
MW: 303.29 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00242764
CAS Number: 78755-81-4
Merck Index: 14,4135

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR