8-(Diethylamino)octyl-3,4,5-trimethoxybenzoate hydrochloride 97%

Supplier: Thermo Scientific Chemicals

J62528-MD J62528-MF
CAAAJ62528-MDEA 1309.71 CAD
CAAAJ62528-MD CAAAJ62528-MF
8-(Diethylamino)octyl-3,4,5-trimethoxybenzoate hydrochloride 97%
8-(Diethylamino)octyl-3,4,5-trimethoxybenzoate hydrochloride

Inhibits intracellular calcium mobilization


Powder


Applications: Inhibits intracellular calcium mobilization

Formula: C₂₂H₃₇NO₅·HCl
Melting Pt: 92…97 °C
Storage Temperature: Ambient
MDL Number: MFCD00012502
CAS Number: 53464-72-5
EINECS: 258-574-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR