(S)-(+)-Camptothecin ≥90%
Supplier: Thermo Scientific Chemicals
|
Danger
|
J62523-06
J62523-MD
J62523-03
CAAAJ62523-06EA
1229.24
CAD
CAAAJ62523-06
CAAAJ62523-MD
CAAAJ62523-03
(S)-(+)-Camptothecin ≥90%
(S)-(+)-Camptothecin
Cytotoxic antitumor agent that inhibits topoisomerase I
CAS No.: 7689-03-4
Molecular Formula: C20H16N2O4
Formula Weight: 348.35
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: Yellow
Melting Point: 260° dec.
Beilstein Reference: 631069
UN No.: 1544
MDL No.: MFCD00081076
Applications: Cytotoxic antitumor agent that inhibits topoisomerase I
Formula:
C₂₀H₁₆N₂O₄ Melting Pt: 260 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00081076 CAS Number: 7689-03-4 UN: 1544 ADR: 6.1,III Merck Index: 14,1735 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...