L(-)-Tryptophan cell culture reagent
Supplier: Thermo Scientific Chemicals
J62508-09
J62508-22
J62508-A1
CAAAJ62508-09EA
73.17
CAD
CAAAJ62508-09
CAAAJ62508-22
CAAAJ62508-A1
L(-)-Tryptophan cell culture reagent
L(-)-Tryptophan
Powder
CAS No.: 73-22-3
Molecular Formula: C11H12N2O2
Formula Weight: 204.23
Physical Form: Powder
Appearance: White to off-white
Melting Point: 280-285°
Beilstein Reference: 86197
Merck Reference: 14,9797
MDL No.: MFCD00064340
Formula:
C₁₁H₁₂N₂O₂ Melting Pt: 280…285 °C Density: 1.34 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00064340 CAS Number: 73-22-3 EINECS: 200-795-6 Merck Index: 14,9797 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...