Tetraethylammonium chloride

Supplier: Thermo Scientific Chemicals
Warning

J62507-14 J62507-22
CAAAJ62507-14EA 69.71 CAD
CAAAJ62507-14 CAAAJ62507-22
Tetraethylammonium chloride
Tetraethylammonium chloride

Phase transfer catalyst


Powder


Phase transfer catalyst

Formula: C₈H₂₀ClN
Melting Pt: 285 °C
Density: 1.08 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011828
CAS Number: 56-34-8
EINECS: 200-267-5
Merck Index: 14,9200

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR