Tetraethylene glycol monooctyl ether

Supplier: Thermo Scientific Chemicals
Warning

J62496-06 J62496-03
CAAAJ62496-06EA 787.05 CAD
CAAAJ62496-06 CAAAJ62496-03
Tetraethylene glycol monooctyl ether
Tetraethylene glycol monooctyl ether

Reagent for solubilizing membrane proteins


Solid


Applications: Reagent for solubilizing membrane proteins

Formula: C₁₆H₃₄O₅
Density: 0.969
Storage Temperature: Freezer
MDL Number: MFCD00043033
CAS Number: 19327-39-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR