Oxytetracycline hydrochloride
Supplier: Thermo Scientific Chemicals
|
Warning
|
J62489-22
J62489-14
CAAAJ62489-22EA
557.91
CAD
CAAAJ62489-22
CAAAJ62489-14
Oxytetracycline hydrochloride
Oxytetracycline hydrochloride
Impairs mitochondrial protein synthesis
Crystalline powder
Impairs mitochondrial protein synthesis
Formula:
C₂₂H₂₅ClN₂O₉ Melting Pt: 180 °C Storage Temperature: Ambient |
MDL Number:
MFCD00135815 CAS Number: 2058-46-0 EINECS: 218-161-2 Merck Index: 14,6976 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...