Nystatin

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Fungicidin, Mycostatin

J62486-06 J62486-09
CAAAJ62486-06EA 147.92 CAD
CAAAJ62486-06 CAAAJ62486-09
Nystatin
Nystatin

An antimicrobial. Increases the permeability of the cell membrane of sensitive fungi by binding to sterols


Fine powder


An antimicrobial. Increases the permeability of the cell membrane of sensitive fungi by binding to sterols

Formula: C₄₇H₇₅NO₁₇
MW: 926.09 g/mol
Melting Pt: 160 °C
Storage Temperature: Refrigerator
MDL Number: MFCD25541651
CAS Number: 1400-61-9
EINECS: 215-749-0
Merck Index: 14,6736

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR