Paraformaldehyde 4% in PBS + Mg + EGTA

Supplier: Thermo Scientific Chemicals
Danger

J62478-AK J62478-AP
CAAAJ62478-AKEA 175.14 CAD
CAAAJ62478-AK CAAAJ62478-AP
Paraformaldehyde 4% in PBS + Mg + EGTA
Paraformaldehyde

For histology.


Liquid


Applications: For histology.

Formula: (CH₂O)n
Storage Temperature: Ambient
MDL Number: MFCD00133991
CAS Number: 30525-89-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR