7-Methoxycoumarin-4-acetic acid ≥99%

Supplier: Thermo Scientific Chemicals

Synonyms: 7-Methoxy-2-oxo-2H-1-benzopyran-4-acetic acid, Mca, 2-(7-Methoxy-2-oxo-2H-chromen-4-yl)acetic acid, (7-Methoxycoumarin-4-yl)acetic acid

J62475-03 J62475-MC J62475-MD
CAAAJ62475-03EA 522.65 CAD
CAAAJ62475-03 CAAAJ62475-MC CAAAJ62475-MD
7-Methoxycoumarin-4-acetic acid ≥99%
7-Methoxycoumarin-4-acetic acid

Precursor to coumarin-type fluorescent reagents


Crystalline solid


Applications: Precursor to coumarin-type fluorescent reagents

Formula: C₁₂H₁₀O₅
MW: 234.207 g/mol
Melting Pt: 193 °C
Flash Pt: 87 °C(189 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009774
CAS Number: 62935-72-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR