Melatonine 99+%

Supplier: Thermo Scientific Chemicals

Synonyms: Melatonin, N-(2-(5-Methoxy-1H-indol-3-yl)ethyl)acetamide

J62452-06 J62452-14
CAAAJ62452-06EA 79.47 CAD
CAAAJ62452-06 CAAAJ62452-14
Melatonine 99+%
Melatonine

A pineal gland hormone associated with sleep regulation. It induces apoptosis


Powder


A pineal gland hormone associated with sleep regulation. It induces apoptosis

Formula: C₁₃H₁₆N₂O₂
MW: 232.28 g/mol
Melting Pt: 116…120 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005655
CAS Number: 73-31-4
EINECS: 200-797-7
Merck Index: 14,5816

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR