KN-62
Supplier: Thermo Scientific Chemicals
J62444-MA
J62444-MCR
CAAAJ62444-MAEA
1846.97
CAD
CAAAJ62444-MA
CAAAJ62444-MCR
KN-62
KN-62
Potently and specifically inhibits calcium/calmodulin kinase II
Powder
Formula:
C₃₈H₃₅N₅O₆S₂ Melting Pt: 65…67 °C Storage Temperature: Freezer |
MDL Number:
MFCD00083180 CAS Number: 127191-97-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...