Penicillin V potassium salt
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Potassium (2S,5R,6R)-3,3-dimethyl-7-oxo-6-(2-phenoxyacetamido)-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate, Penicillin V Potassium
J62442-09
J62442-14
J62442-22
CAAAJ62442-09EA
119.26
CAD
CAAAJ62442-09
CAAAJ62442-14
CAAAJ62442-22
Penicillin V potassium salt
Penicillin V potassium salt
Inhibits bacterial cell wall synthesis
White to off-white powder
Formula:
C₁₆H₁₇KN₂O₅S MW: 388.48 g/mol Melting Pt: 197…202 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00051771 CAS Number: 132-98-9 EINECS: 205-086-5 Merck Index: 14,7100 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...