Potassium chloride 1 M in aqueous solution RNase-free

Supplier: Thermo Scientific Chemicals

J62422-AE J62422-AK
AAJ62422-AEEA 148.19 CAD
AAJ62422-AE CAAAJ62422-AK CAAAJ62422-EQE
Potassium chloride 1 M in aqueous solution RNase-free
Potassium chloride

Liquid


CAS No.: 7447-40-7 Molecular Formula: ClK Formula Weight: 74.55 Physical Form: Liquid MDL No.: MFCD00011360

Formula: KCl
Density: 1.13 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011360
CAS Number: 7447-40-7
EINECS: 231-211-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR