Triolein ≥75%, tech.
Supplier: Thermo Scientific Chemicals
J62419-14
J62419-22
CAAAJ62419-14EA
270.94
CAD
CAAAJ62419-14
CAAAJ62419-22
Triolein ≥75%, tech.
Triolein
Liquid
Purity: 75%
CAS No.: 122-32-7
Molecular Formula: C57H104O6
Formula Weight: 885.45
Storage Temperature: -30°C to -10°C
Flash Point: 330°(626°F)
Physical Form: Liquid
Density: 0.91
Refractive Index: 1.468
Appearance: Clear pale yellow
Boiling Point: 235-240°/18mm
Solubility: Soluble in chlorofom, ether, and carbon tetrachloride
Beilstein Reference: 1718692
MDL No.: MFCD00137563
Formula:
C₅₇H₁₀₄O₆ Boiling Pt: 235…240 °C/18 mm Density: 0.91 Flash Pt: 330 °C(626 °F) Storage Temperature: Freezer |
MDL Number:
MFCD00137563 CAS Number: 122-32-7 EINECS: 204-534-7 Merck Index: 14,9732 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...