Magnesium chloride 1 M in aqueous solution

Supplier: Thermo Scientific Chemicals

J62411-AD J62411-AE
CAAAJ62411-ADEA 100.03 CAD
CAAAJ62411-AD CAAAJ62411-AE
Magnesium chloride 1 M in aqueous solution
Magnesium chloride

Liquid


CAS No.: 7786-30-3 Molecular Formula: Cl2Mg Formula Weight: 95.22 Physical Form: Liquid MDL No.: MFCD00011106

Formula: MgCl₂
Storage Temperature: Ambient
MDL Number: MFCD00011106
CAS Number: 7786-30-3
EINECS: 232-094-6
Merck Index: 13,05684

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR