AMPSO 98+%

Supplier: Thermo Scientific Chemicals

J62378-14 J62378-22
CAAAJ62378-14EA 85.76 CAD
CAAAJ62378-14 CAAAJ62378-22
AMPSO 98+%
AMPSO

Useful as a buffer


Crystals


Useful as a buffer

Formula: C₇H₁₇NO₅S
Melting Pt: 215…225 °C
Storage Temperature: Ambient
MDL Number: MFCD00041777
CAS Number: 68399-79-1
EINECS: 269-991-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR