Irinotecan hydrochloride trihydrate 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: CPT-11, Irinotecan HCl Trihydrate, (S)-4,11-Diethyl-4-hydroxy-3,14-dioxo-3,4,12,14-tetrahydro-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinolin-9-yl [1,4'-bipiperidine]-1'-carboxylate hydrochloride trihydrate

J62370-MC J62370-MD
CAAAJ62370-MCEA 383.32 CAD
CAAAJ62370-MC CAAAJ62370-MD
Irinotecan hydrochloride trihydrate 99%
Irinotecan hydrochloride trihydrate

Topoisomerase I inhibitor


Crystalline powder


Applications: Topoisomerase I inhibitor

Formula: C₃₃H₃₈N₄O₆·HCl·3H₂O
MW: 677.19 g/mol (623.15 anhy)
Melting Pt: 256 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01765731
CAS Number: 136572-09-3
Merck Index: 14,5091

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR