Camostat methanesulfate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Camostat Methanesulfonate, 4-(2-(2-(Dimethylamino)-2-oxoethoxy)-2-oxoethyl)phenyl 4-guanidinobenzoate methanesulfonate, Camostat Mesylate, Camostat Methanesulphonate, 4-(2-(2-(Dimethylamino)-2-oxoethoxy)-2-oxoethyl)phenyl 4-guanidinobenzoate methanesulphonate

J62362-MC J62362-ME
CAAAJ62362-MCEA 824.3 CAD
CAAAJ62362-MC CAAAJ62362-ME
Camostat methanesulfate 98%
Camostat methanesulfate

An orally active synthetic serine protease inhibitor


Powder


Applications: An orally active synthetic serine protease inhibitor

Formula: C₂₀H₂₂N₄O₅·CH₃SO₃H
MW: 494.52 g/mol
Melting Pt: 164…168 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00941410
CAS Number: 59721-29-8
UN: 3077
ADR: "9",III
Merck Index: 14,1728

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR