BIS-TRIS Propane, 0.2 M Buffer Solution, pH 6.5, Thermo Scientific
Supplier: Thermo Scientific Chemicals
J62354-AE
J62354-AK
CAAAJ62354-AEEA
175.14
CAD
CAAAJ62354-AE
CAAAJ62354-AK
BIS-TRIS Propane, 0.2 M Buffer Solution, pH 6.5, Thermo Scientific
Buffers
Biological Buffers
Liquid
CAS No.: 64431-96-5
Physical Form: Liquid
MDL No.: MFCD00004689
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...