Phosphate, 0.5 M Buffer Solution, pH 9.0, Thermo Scientific
Supplier: Thermo Scientific Chemicals
J62345-AK
J62345-AP
CAAAJ62345-AKEA
112.45
CAD
CAAAJ62345-AK
CAAAJ62345-AP
Phosphate, 0.5 M Buffer Solution, pH 9.0, Thermo Scientific
Buffers
Biological Buffers
Liquid
CAS No.: 7558-80-7
Physical Form: Liquid
MDL No.: MFCD00003527
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...