β-Nicotinamide adenine dinucleotid, oxidised form (NAD, oxidised form) 97%
Supplier: Thermo Scientific Chemicals
Synonyms:
Nadide, NAD+, Nicotinamide adenine dinucleotide, oxidized
J62337-03
J62337-06
J62337-14
CAAAJ62337-03EA
111.97
CAD
CAAAJ62337-03
CAAAJ62337-06
CAAAJ62337-14
β-Nicotinamide adenine dinucleotid, oxidised form (NAD, oxidised form) 97%
β-Nicotinamide adenine dinucleotide (NAD, oxidized form)
Powder
Purity: 98+%
CAS No.: 53-84-9
Molecular Formula: C21H27N7O14P2
Formula Weight: 663.43
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White
Solubility: Soluble in water at 50mg/ml
Beilstein Reference: 3584133
MDL No.: MFCD00036253
Formula:
C₂₁H₂₇N₇O₁₄P₂ MW: 663.43 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00036253 CAS Number: 53-84-9 EINECS: 200-184-4 Merck Index: 14,6344 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...