Maprotiline hydrochloride 99%

Supplier: Thermo Scientific Chemicals
Warning

J62321-03 J62321-06
CAAAJ62321-03EA 219.29 CAD
CAAAJ62321-03 CAAAJ62321-06
Maprotiline hydrochloride 99%
Maprotiline hydrochloride

Selective norepinephrine uptake inhibitor


Crystalline powder


Applications: Selective norepinephrine uptake inhibitor

Formula: C₂₀H₂₃N
Melting Pt: 230…232 °C
Storage Temperature: Ambient
MDL Number: MFCD00079464
CAS Number: 10347-81-6
EINECS: 233-758-8
Merck Index: 14,5748

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR