Azathioprine

Supplier: Thermo Scientific Chemicals
Danger

J62314-03 J62314-06 J62314-09
CAAAJ62314-03EA 241.53 CAD
CAAAJ62314-03 CAAAJ62314-06 CAAAJ62314-09
Azathioprine
Azathioprine

An immunosuppressant. A purine synthesis inhibitor which inhibits the proliferation of cells


Crystalline powder


Applications: An immunosuppressant. A purine synthesis inhibitor which inhibits the proliferation of cells

Formula: C₉H₇N₇O₂S
Storage Temperature: Freezer
MDL Number: MFCD00069203
CAS Number: 446-86-6
EINECS: 207-175-4
Merck Index: 14,902

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR