L-Cystine disodium salt ≥98%
Supplier: Thermo Scientific Chemicals
Synonyms:
Sodium (2R,2'R)-3,3'-disulphanediylbis(2-aminopropanoate), Sodium (2R,2'R)-3,3'-disulfanediylbis(2-aminopropanoate)
J62310-30
J62310-A1
CAAAJ62310-30EA
278.32
CAD
CAAAJ62310-30
CAAAJ62310-A1
L-Cystine disodium salt ≥98%
L-Cystine disodium salt
Powder
Purity: 98+%
CAS No.: 64704-23-0
Molecular Formula: C6H10N2Na2O4S2
Formula Weight: 284.30
Physical Form: Powder
Appearance: Pale brown
Optical Rotation: +170° (c=2 in 1N HCl)
MDL No.: MFCD00037206
Formula:
C₆H₁₀N₂Na₂O₄S₂ MW: 284.30 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00037206 CAS Number: 64704-23-0 EINECS: 265-025-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...