Cilostazol 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 6-(4-(1-Cyclohexyl-1H-tetrazol-5-yl)butoxy)-3,4-dihydroquinolin-2(1H)-one, 6-[4-(1-Cyclohexyl-1H-tetrazol-5-yl)butoxy]-3,4-dihydro-2(1H)-quinolinone

J62301-MA J62301-MF
CAAAJ62301-MAEA 335.94 CAD
CAAAJ62301-MA CAAAJ62301-MF
Cilostazol 98%
Cilostazol

An inhibitor of phosphodiesterase III


Solid


Applications: An inhibitor of phosphodiesterase III

Formula: C₂₀H₂₇N₅O₂
MW: 369.47 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00866780
CAS Number: 73963-72-1
Merck Index: 14,2277

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR