Triton® X-100 lysis buffer, pH 7.4, Thermo Scientific
Supplier: Thermo Scientific Chemicals
J62289-AK
J62289-AP
CAAAJ62289-AKEA
121.41
CAD
CAAAJ62289-AK
CAAAJ62289-AP
Triton® X-100 lysis buffer, pH 7.4, Thermo Scientific
Buffers
Extraction Buffers
Liquid
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Appearance: Colorless
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...