TE Buffer, pH 7.6, RNAse Free, Thermo Scientific
Supplier: Thermo Scientific Chemicals
J62285-AK
J62285-EQE
CAAAJ62285-AKEA
225.1
CAD
CAAAJ62285-AK
CAAAJ62285-EQE
TE Buffer, pH 7.6, RNAse Free, Thermo Scientific
Buffers
Biological Buffers
Liquid
Physical Form: Liquid
MDL No.: MFCD00236359
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...