Human beta-Casomorphin
Supplier: Thermo Scientific Chemicals
J62271-LB0
J62271-MB
CAAAJ62271-LB0EA
172.8
CAD
CAAAJ62271-LB0
CAAAJ62271-MB
Human beta-Casomorphin
Proteins and Peptides
An opioid peptide.
Purity: 95+%
CAS No.: 102029-74-3
Molecular Formula: C44H61N7O11
Formula Weight: 864.01
Storage Temperature: -30°C to -10°C
Physical Form: Powder
MDL No.: MFCD00076324
An opioid peptide
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...