Sudan black B

Supplier: Thermo Scientific Chemicals

J62268-09 J62268-14 J62268-22
CAAAJ62268-09EA 83.9 CAD
CAAAJ62268-09 CAAAJ62268-14 CAAAJ62268-22
Sudan black B
Sudan black B

Histochemical stain for chromosomes


Powder


Histochemical stain for chromosomes

Formula: C₂₉H₂₄N₆
Melting Pt: 118…126 °C
Storage Temperature: Ambient
MDL Number: MFCD00006919
CAS Number: 4197-25-5
EINECS: 224-087-1
Merck Index: 14,8885

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR