5-Iodouridine 96%

Supplier: Thermo Scientific Chemicals

Synonyms: 1-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-iodopyrimidine-2,4(1H,3H)-dione

J62259-03 J62259-06
CAAAJ62259-03EA 239.91 CAD
CAAAJ62259-03 CAAAJ62259-06
5-Iodouridine 96%
5-Iodouridine

Powder


Purity: 96% CAS No.: 1024-99-3 Molecular Formula: C9H11IN2O6 Formula Weight: 370.10 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White Beilstein Reference: 33665 MDL No.: MFCD00006532

Formula: C₉H₁₁IN₂O₆
MW: 370.10 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00006532
CAS Number: 1024-99-3
EINECS: 213-833-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR