Pirenzepine dihydrochloride 99%

Supplier: Thermo Scientific Chemicals

J62252-03 J62252-MC
CAAAJ62252-03EA 358.64 CAD
CAAAJ62252-03 CAAAJ62252-06 CAAAJ62252-MC
Pirenzepine dihydrochloride 99%
Pirenzepine dihydrochloride

A M1 muscarinic receptor antagonist


Powder


Applications: A M1 muscarinic receptor antagonist

Formula: C₁₉H₂₁N₅O₂·2HCl
Melting Pt: 192…198 °C
Storage Temperature: Ambient
MDL Number: MFCD00055214
CAS Number: 29868-97-1
EINECS: 249-907-5
Merck Index: 14,7491

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR