1H-[1,2,4]Oxadiazolo[4,3-a]quinoxalin-1-one ≥99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: ODQ

J62250-MA J62250-MF
CAAAJ62250-MAEA 237.52 CAD
CAAAJ62250-MA CAAAJ62250-MF
1H-[1,2,4]Oxadiazolo[4,3-a]quinoxalin-1-one ≥99%
1H-[1,2,4]Oxadiazolo[4,3-a]quinoxalin-1-one

Selective inhibitor of nitric oxide-sensitive guanylyl cyclase


Powder


Applications: Selective inhibitor of nitric oxide-sensitive guanylyl cyclase

Formula: C₉H₅N₃O₂
MW: 187.16 g/mol
Melting Pt: 160…170 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00792620
CAS Number: 41443-28-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR