Cytidine-5'-triphosphoric acid disodium salt (CTP-Na2) 97%
Supplier: Thermo Scientific Chemicals
Synonyms:
Cytidine-5'-triphosphate disodium, Sodium ((2R,3S,4R,5R)-5-(4-amino-2-oxopyrimidin-1(2H)-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl dihydrogentriphosphate
J62238-03
J62238-04
J62238-ME
CAAAJ62238-03EA
538.67
CAD
CAAAJ62238-03
CAAAJ62238-04
CAAAJ62238-ME
Cytidine-5'-triphosphoric acid disodium salt (CTP-Na2) 97%
Cytidine-5'-triphosphoric acid disodium salt (CTP-Na2)
Powder
Purity: 97+%
CAS No.: 36051-68-0
Molecular Formula: C9H14N3Na2O14P3
Formula Weight: 527.12
Flash Point: 113°(235°F)
Physical Form: Powder
MDL No.: MFCD00078193
Formula:
C₉H₁₄N₃Na₂O₁₄P₃ MW: 527.12 g/mol Flash Pt: 113 °C(235 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00078193 CAS Number: 36051-68-0 EINECS: 252-849-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...